This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02809378_m1
See other SLC2A3 GE Assays ›
Gene Symbol:
SLC2A3
Gene Name:
solute carrier family 2 (facilitated glucose transporter), member 3
Gene Aliases:
-
Chromosome Location:
Chr.11: 11576181 - 11592299 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02809378_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC2A3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001261533.1 NP_001248462.1 1296 99
GenBank mRNA JU322631.1 - 1255 99
JU322632.1 - 1255 99
JU322633.1 - 1255 99
JU322635.1 - 1255 99
JU322636.1 - 1255 99
JU332195.1 - 1255 99
JU332196.1 - 1255 99
JU332197.1 - 1255 99
JU332198.1 - 1255 99
JU332199.1 - 1255 99
JU470910.1 - 1296 99
JU470911.1 - 1296 99
JU470912.1 - 1296 99
JU470913.1 - 1296 99
JU470914.1 - 1296 99
JU470915.1 - 1296 99
JU470916.1 - 1296 99
JU470917.1 - 1296 99
JU470918.1 - 1296 99
JU470919.1 - 1296 99

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header