This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02801773_m1
See other S100A6 GE Assays ›
Gene Symbol:
S100A6
Gene Name:
S100 calcium binding protein A6
Gene Aliases:
EGK_01326
Chromosome Location:
Chr.1: 97533171 - 97534795 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02801773_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100A6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001261799.1 NP_001248728.1 275 68
XM_028845294.1 XP_028701127.1 264 68
GenBank mRNA JU320264.1 - 259 68
JU473185.1 - 275 68
JV658295.1 - 284 68

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length less than or equal to 70 3' Most Macaca mulatta

Panther Classification:

Molecular Function -

calmodulin-related

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0