Assay Details
Gene Symbol: CRYL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266898.1 | NP_001253827.1 | 796 | 116 | ||
XM_015120672.2 | XP_014976158.1 | 668 | 116 | |||
XM_028837116.1 | XP_028692949.1 | 830 | 116 | |||
XM_028837117.1 | XP_028692950.1 | 830 | 116 | |||
XM_028837118.1 | XP_028692951.1 | 688 | 116 | |||
XM_028837119.1 | XP_028692952.1 | 669 | 116 | |||
XM_028837120.1 | XP_028692953.1 | 820 | 116 | |||
XM_028837122.1 | XP_028692955.1 | 807 | 116 | |||
XM_028837123.1 | XP_028692956.1 | 720 | 116 | |||
XM_028837124.1 | XP_028692957.1 | 926 | 116 | |||
GenBank mRNA | JU334776.1 | - | 810 | 116 | ||
JU475818.1 | - | 748 | 116 | |||
JV047242.1 | - | 782 | 116 | |||
JV565133.1 | - | 796 | 116 | |||
JV716257.1 | - | 774 | 116 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
