This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02796810_m1
See other RCAN1 GE Assays ›
Gene Symbol:
RCAN1
Gene Name:
regulator of calcineurin 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 11954956 - 12053394 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02796810_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RCAN1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001088613.4 XP_001088613.1 604 88
XM_015132920.2 XP_014988406.1 606 88
XM_015132921.2 XP_014988407.1 1087 88
XM_015132922.2 XP_014988408.1 1087 88
XM_028845331.1 XP_028701164.1 551 88
GenBank mRNA JU322262.1 - 364 88
JU326676.1 - 373 88
JV708647.1 - 367 88

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_028845331.1 transcripts.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

intercellular signal molecule

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0