This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02792531_m1
See other NPC2 GE Assays ›
Gene Symbol:
NPC2
Gene Name:
NPC intracellular cholesterol transporter 2
Gene Aliases:
-
Chromosome Location:
Chr.7: 136591300 - 136605436 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02792531_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NPC2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001266760.1 NP_001253689.1 290 81
XM_015144113.2 XP_014999599.1 269 81
XM_028850263.1 XP_028706096.1 325 81
XM_028850264.1 XP_028706097.1 268 81
GenBank mRNA JU320610.1 - 272 81
JU474016.1 - 263 81
JV045946.1 - 290 81
JV640371.1 - 262 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0