Assay Details
Gene Symbol: TP53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047151.2 | NP_001040616.1 | 1090 | 81 | ||
XM_028835713.1 | XP_028691546.1 | 1277 | 81 | |||
GenBank mRNA | HM104190.1 | - | 1074 | 81 | ||
HM104191.1 | - | 1074 | 81 | |||
JU472158.1 | - | 1162 | 81 | |||
JU472159.1 | - | 1162 | 81 | |||
JU472160.1 | - | 1162 | 81 | |||
JV044751.1 | - | 1133 | 81 | |||
L20442.1 | - | 1090 | 81 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
