This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02789143_m1
See other CNP GE Assays ›
Gene Symbol:
CNP
Gene Name:
2',3'-cyclic nucleotide 3' phosphodiesterase
Gene Aliases:
-
Chromosome Location:
Chr.16: 53090947 - 53102448 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02789143_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CNP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001035528.3 NP_001030605.2 969 62
XM_028835460.1 XP_028691293.1 1219 62
XM_028835461.1 XP_028691294.1 958 62
GenBank mRNA DQ188084.1 - 822 62
JU474298.1 - 925 62
JU474299.1 - 925 62
JU474300.1 - 925 62
JU474301.1 - 925 62
JU474302.1 - 925 62
JV046181.1 - 932 62
JV046182.1 - 932 62
JV601055.1 - 969 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

phosphodiesterase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0