This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02789112_m1
See other PDIA3 GE Assays ›
Gene Symbol:
PDIA3
Gene Name:
protein disulfide isomerase family A member 3
Gene Aliases:
GRP58
Chromosome Location:
Chr.7: 20228526 - 20253916 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02789112_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PDIA3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001195716.1 NP_001182645.1 1124 107
GenBank mRNA CN647099.1 - 612 107
DQ148150.1 - 380 107
JU321862.1 - 1175 107
JU472011.1 - 1183 107
JV043339.1 - 1186 107

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

chaperone

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0