This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02787591_m1
See other CRHR1 GE Assays ›
Gene Symbol:
CRHR1
Gene Name:
corticotropin releasing hormone receptor 1
Gene Aliases:
CRF1
Chromosome Location:
Chr.16: 57581792 - 57638936 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02787591_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CRHR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001032803.1 NP_001027975.1 807 58
XM_015119719.2 XP_014975205.1 1070 58
XM_015119720.2 XP_014975206.1 852 58
XM_028835443.1 XP_028691276.1 1710 58
XM_028835444.1 XP_028691277.1 950 58
XM_028835445.1 XP_028691278.1 972 58
XM_028835446.1 XP_028691279.1 1157 58
GenBank mRNA AB078141.1 - 807 58

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Macaca mulatta Inventoried

Panther Classification:

Molecular Function -

transmembrane signal receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0