This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh01567292_g1
See other NRBF2 GE Assays ›
Gene Symbol:
NRBF2
Gene Name:
nuclear receptor binding factor 2
Gene Aliases:
EGK_19822
Chromosome Location:
Chr.9: 74826037 - 74848118 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh01567292_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NRBF2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001261731.1 NP_001248660.1 192 81
GenBank mRNA JU334227.1 - 178 81
JU475342.1 - 237 81
JV046890.1 - 192 81
JV046891.1 - 192 81
JV635601.1 - 238 81
JV703513.1 - 182 81

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001261731.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 3' Most Macaca mulatta

Panther Classification:

Molecular Function -

transcription cofactor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0