This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh01121703_m1
See other TSG101 GE Assays ›
Gene Symbol:
TSG101
Gene Name:
tumor susceptibility 101
Gene Aliases:
EGK_06430
Chromosome Location:
Chr.14: 47040967 - 47092442 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh01121703_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TSG101
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001195481.1 NP_001182410.1 278 88
XM_015114617.2 XP_014970103.1 225 88
GenBank mRNA CN644066.1 - 278 88
DQ902357.1 - 100 88
DQ902363.1 - 101 88
DQ902365.1 - 107 88
DQ902366.1 - 107 88
DQ902367.1 - 112 88
DQ902368.1 - 104 88
DQ902369.1 - 60 88
DQ902370.1 - 52 88
DQ902372.1 - 61 88
DQ902373.1 - 124 88
DQ902375.1 - 109 88
DQ902376.1 - 111 88
DQ902377.1 - 109 88
DQ902378.1 - 127 88
DQ902379.1 - 54 88
DQ902380.1 - 74 88
DQ902381.1 - 53 88
DQ902383.1 - 102 88
DQ902384.1 - 60 88
DQ902385.1 - 57 88
DQ902386.1 - 60 88
DQ902387.1 - 69 88
DQ902390.1 - 61 88
DQ902391.1 - 59 88
DQ902392.1 - 105 88
DQ902395.1 - 58 88
DQ902398.1 - 61 88
DQ902399.1 - 106 88
JU336180.1 - 312 88
JU477035.1 - 208 88
JV048128.1 - 210 88

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the XM_015114617.2 transcripts.

Set Membership:

Amplicon length between 86 and 100 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0