This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh01105502_m1
See other CTDSP1 GE Assays ›
Gene Symbol:
CTDSP1
Gene Name:
CTD small phosphatase 1
Gene Aliases:
-
Chromosome Location:
Chr.12: 105816269 - 105823776 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh01105502_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CTDSP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001193873.1 NP_001180802.1 716 86
XM_015110974.2 XP_014966460.2 547 86
XM_015110975.2 XP_014966461.2 544 86
XM_015110976.1 XP_014966462.1 708 86
XM_015110977.1 XP_014966463.1 392 86
GenBank mRNA JU335713.1 - 472 86
JU476597.1 - 470 86
JV047795.1 - 453 86
JV589402.1 - 447 86
JV600818.1 - 464 86
JV667440.1 - 460 86

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0