This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh01103386_m1
See other SNCA GE Assays ›
Gene Symbol:
SNCA
Gene Name:
synuclein alpha
Gene Aliases:
EGK_15919
Chromosome Location:
Chr.5: 88118358 - 88230916 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh01103386_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SNCA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015138783.2 XP_014994269.1 980 97
XM_015138784.2 XP_014994270.1 959 97
XM_015138785.2 XP_014994271.1 1235 97
XM_015138786.2 XP_014994272.1 514 97
XM_015138787.2 XP_014994273.1 514 97
XM_015138788.2 XP_014994274.1 525 97
XM_015138789.2 XP_014994275.1 1131 97
XM_015138790.2 XP_014994276.1 559 97
XM_015138791.2 XP_014994277.1 438 97
XM_028848623.1 XP_028704456.1 542 97
XM_028848624.1 XP_028704457.1 531 97
XM_028848625.1 XP_028704458.1 525 97
GenBank mRNA JU321172.1 - 582 97
JU336039.1 - 662 97
JV045139.1 - 525 97

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta Inventoried

Panther Classification:

Molecular Function -

membrane trafficking regulatory protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0