This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh01097019_m1
See other PPP2R3A GE Assays ›
Gene Symbol:
PPP2R3A
Gene Name:
protein phosphatase 2 regulatory subunit B''alpha
Gene Aliases:
EGK_12083
Chromosome Location:
Chr.2: 38124864 - 38318602 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh01097019_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP2R3A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001257608.1 NP_001244537.1 2947 85
XM_015132061.2 XP_014987547.1 3209 85
XM_015132062.2 XP_014987548.1 3336 85
XM_015132063.2 XP_014987549.1 956 85
XM_028843433.1 XP_028699266.1 3197 85
XM_028843434.1 XP_028699267.1 3155 85
GenBank mRNA JU322700.1 - 2947 85
JU322701.1 - 2947 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

protein phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0