This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh01093120_m1
See other CTNNA2 GE Assays ›
Gene Symbol:
CTNNA2
Gene Name:
catenin alpha 2
Gene Aliases:
EGK_05497
Chromosome Location:
Chr.13: 27964763 - 29138405 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh01093120_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CTNNA2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001266343.1 NP_001253272.1 1528 57
XM_015112724.2 XP_014968210.1 1662 57
XM_015112725.2 XP_014968211.1 1635 57
XM_015112726.2 XP_014968212.1 593 57
XM_028831580.1 XP_028687413.1 609 57
XM_028831581.1 XP_028687414.1 420 57
GenBank mRNA JU320100.1 - 1528 57
JU320101.1 - 1528 57
JV534453.1 - 1490 57
JV606881.1 - 1616 57

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

non-motor actin binding protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0