This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh01086495_m1
See other LOC106994601 GE Assays ›
Gene Symbol:
LOC106994601 SPPL2B
Gene Name:
signal peptide peptidase-like 2B
signal peptide peptidase like 2B
Gene Aliases:
SPPL2B
-
Chromosome Location:
Chr.19: 66231 - 84344 on Build Mmul_10
Chromosome Location:
Chr.19: 2177859 - 2199242 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
-
Assay ID Rh01086495_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC106994601
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_028842813.1 XP_028698646.1 620 70
XM_028842814.1 XP_028698647.1 617 70
XM_028842815.1 XP_028698648.1 469 70
XM_028842816.1 XP_028698649.1 755 70
GenBank mRNA JU325140.1 - 815 70
JV656405.1 - 823 70
JV656407.1 - 823 70
Gene Symbol: SPPL2B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_028838678.1 XP_028694511.1 987 70
GenBank mRNA JV513559.1 - 800 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Macaca mulatta

Panther Classification:

Molecular Function -

aspartic protease

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0