This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh01074927_m1
See other GRB2 GE Assays ›
Gene Symbol:
GRB2
Gene Name:
growth factor receptor bound protein 2
Gene Aliases:
EGK_08977
Chromosome Location:
Chr.16: 71685591 - 71773982 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh01074927_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GRB2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015120312.2 XP_014975798.1 487 74
XM_028836787.1 XP_028692620.1 339 74
XM_028836788.1 XP_028692621.1 348 74
GenBank mRNA JU334126.1 - 329 74
JU334127.1 - 329 74
JU334128.1 - 329 74
JU475265.1 - 351 74
JV046826.1 - 339 74
JV046827.1 - 339 74
JV046828.1 - 339 74
JV046829.1 - 339 74
JV046830.1 - 339 74
JV046831.1 - 339 74

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0