Assay Details
Gene Symbol: RNPEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001107559.3 | XP_001107559.1 | 1146 | 67 | ||
XM_015120255.2 | XP_014975741.1 | 779 | 67 | |||
XM_028852048.1 | XP_028707881.1 | 997 | 67 | |||
XM_028852056.1 | XP_028707889.1 | 571 | 67 | |||
XM_028852067.1 | XP_028707900.1 | 936 | 67 | |||
GenBank mRNA | JU327336.1 | - | 812 | 67 | ||
JU472455.1 | - | 1101 | 67 | |||
JV638719.1 | - | 1107 | 67 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
