Assay Details
Gene Symbol: N4BP2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015120852.2 | XP_014976338.2 | 1763 | 95 | ||
XM_015120853.2 | XP_014976339.2 | 1733 | 95 | |||
XM_028837650.1 | XP_028693483.1 | 431 | 95 | |||
XM_028837652.1 | XP_028693485.1 | 492 | 95 | |||
XM_028837653.1 | XP_028693486.1 | 368 | 95 | |||
GenBank mRNA | JU323696.1 | - | 1742 | 95 | ||
JV705233.1 | - | 1739 | 95 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
