Assay Details
Gene Symbol: TIA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261758.1 | NP_001248687.1 | 1261 | 57 | ||
XM_015112521.2 | XP_014968007.1 | 1397 | 57 | |||
XM_015112524.2 | XP_014968010.1 | 1367 | 57 | |||
XM_028831466.1 | XP_028687299.1 | 1364 | 57 | |||
XM_028831467.1 | XP_028687300.1 | 1043 | 57 | |||
XM_028831468.1 | XP_028687301.1 | 1342 | 57 | |||
XM_028831469.1 | XP_028687302.1 | 1940 | 57 | |||
XM_028831470.1 | XP_028687303.1 | 861 | 57 | |||
XM_028831471.1 | XP_028687304.1 | 1255 | 57 | |||
XM_028831472.1 | XP_028687305.1 | 658 | 57 | |||
GenBank mRNA | JU336134.1 | - | 1253 | 57 | ||
JU336135.1 | - | 1220 | 57 | |||
JU476996.1 | - | 1248 | 57 | |||
JU476997.1 | - | 1215 | 57 | |||
JV048088.1 | - | 1261 | 57 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
