Assay Details
Gene Symbol: CAB39L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015121076.2 | XP_014976562.1 | 908 | 102 | ||
XM_015121077.2 | XP_014976563.1 | 846 | 102 | |||
XM_028837175.1 | XP_028693008.1 | 650 | 102 | |||
XM_028837176.1 | XP_028693009.1 | 645 | 102 | |||
XM_028837177.1 | XP_028693010.1 | 530 | 102 | |||
XM_028837178.1 | XP_028693011.1 | 556 | 102 | |||
XM_028837179.1 | XP_028693012.1 | 895 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
