This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh01026927_m1
See other CCN2 GE Assays ›
Gene Symbol:
CCN2
Gene Name:
cellular communication network factor 2
Gene Aliases:
CTGF
Chromosome Location:
Chr.4: 35499610 - 35502866 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh01026927_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CCN2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015137431.2 XP_014992917.2 806 70
XM_028847128.1 XP_028702961.1 855 70
GenBank mRNA JU333088.1 - 560 70
JU333089.1 - 560 70
JV639404.1 - 752 70

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta Inventoried

Panther Classification:

Molecular Function -

growth factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0