This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh01025495_m1
See other ILDR2 GE Assays ›
Gene Symbol:
ILDR2
Gene Name:
immunoglobulin like domain containing receptor 2
Gene Aliases:
-
Chromosome Location:
Chr.1: 24268956 - 24330524 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh01025495_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ILDR2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015125393.2 XP_014980879.1 428 63
XM_015125406.2 XP_014980892.1 495 63
XM_028850028.1 XP_028705861.1 494 63
XM_028850036.1 XP_028705869.1 494 63
XM_028850047.1 XP_028705880.1 496 63
XM_028850053.1 XP_028705886.1 495 63
XM_028850055.1 XP_028705888.1 494 63
GenBank mRNA JU324696.1 - 421 63
JV544383.1 - 438 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

defense/immunity protein immunoglobulin receptor superfamily

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0