This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh01012760_m1
See other SLC25A23 GE Assays ›
Gene Symbol:
SLC25A23
Gene Name:
solute carrier family 25 member 23
Gene Aliases:
-
Chromosome Location:
Chr.19: 6370274 - 6396645 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh01012760_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC25A23
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001257915.1 NP_001244844.1 769 58
XM_015122625.2 XP_014978111.1 806 58
XM_015122626.2 XP_014978112.1 806 58
XM_028838360.1 XP_028694193.1 806 58
XM_028838361.1 XP_028694194.1 806 58
XM_028838362.1 XP_028694195.1 806 58
GenBank mRNA JU320166.1 - 769 58
JU320167.1 - 769 58

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

mitochondrial carrier protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0