Assay Details
Gene Symbol: PCBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260867.1 | NP_001247796.1 | 353 | 56 | ||
XM_015130550.2 | XP_014986036.1 | 377 | 56 | |||
XM_015130551.2 | XP_014986037.1 | 1157 | 56 | |||
XM_015130552.2 | XP_014986038.1 | 1023 | 56 | |||
XM_015130553.2 | XP_014986039.1 | 3129 | 56 | |||
XM_015130555.2 | XP_014986041.1 | 989 | 56 | |||
XM_015130556.2 | XP_014986042.1 | 343 | 56 | |||
XM_028843155.1 | XP_028698988.1 | 1643 | 56 | |||
GenBank mRNA | JU322285.1 | - | 292 | 56 | ||
JU322286.1 | - | 440 | 56 | |||
JU473763.1 | - | 204 | 56 | |||
JV045086.1 | - | 353 | 56 | |||
JV622852.1 | - | 321 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
