This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh00855984_m1
See other PRSS53 GE Assays ›
Gene Symbol:
PRSS53
Gene Name:
serine protease 53
Gene Aliases:
-
Chromosome Location:
Chr.20: 28508318 - 28515419 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh00855984_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRSS53
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001112012.4 XP_001112012.1 1953 113
XM_015126005.2 XP_014981491.1 2097 113
XM_015126006.2 XP_014981492.1 2095 113
XM_015126007.2 XP_014981493.1 1981 113
XM_015126008.2 XP_014981494.1 1956 113
XM_028840735.1 XP_028696568.1 1840 113
XM_028840736.1 XP_028696569.1 2097 113
GenBank mRNA JV615199.1 - 635 113

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

protease serine protease protein modifying enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0