Assay Details
Gene Symbol: POGLUT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001104429.4 | XP_001104429.1 | 990 | 84 | ||
XM_015115618.2 | XP_014971104.1 | 949 | 84 | |||
XM_028833944.1 | XP_028689777.1 | 904 | 84 |
