This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh00427620_m1
See other TBP GE Assays ›
Gene Symbol:
TBP
Gene Name:
TATA-box binding protein
Gene Aliases:
-
Chromosome Location:
Chr.4: 83600 - 102885 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh00427620_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TBP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001085368.4 XP_001085368.1 970 91
XM_015137757.2 XP_014993243.1 812 91
XM_015137759.2 XP_014993245.1 771 91
XM_015137760.2 XP_014993246.1 746 91
XM_015137762.2 XP_014993248.1 685 91
XM_028846940.1 XP_028702773.1 750 91
XM_028846941.1 XP_028702774.1 758 91
GenBank mRNA JV722073.1 - 225 91

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 86 and 100 Endogenous Controls Recommended Assay Macaca mulatta Inventoried

Panther Classification:

Molecular Function -

general transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0