This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh00355361_m1
See other YARS1 GE Assays ›
Gene Symbol:
YARS1
Gene Name:
tyrosyl-tRNA synthetase 1
Gene Aliases:
EGK_00518, YARS
Chromosome Location:
Chr.1: 191828845 - 191873228 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh00355361_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: YARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015134550.2 XP_014990036.1 1595 69
XM_028836407.1 XP_028692240.1 940 69
XM_028836408.1 XP_028692241.1 1115 69
GenBank mRNA JU321904.1 - 838 69
JU471219.1 - 841 69
JV045536.1 - 844 69
JV566762.1 - 872 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

translational protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0