Assay Details
Gene Symbol: LOC4342472 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791517.2 | XP_015647003.1 | 11 - 12 | 1372 | 67 | |
GenBank mRNA | AK111557.1 | - | 11 - 12 | 1308 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
