This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03624200_g1
See other LOC4331069 GE Assays ›
Gene Symbol:
LOC4331069 LOC4340405
Gene Name:
mRNA-decapping enzyme subunit 2
polyadenylate-binding protein-interacting protein 7
Gene Aliases:
OJ1111_C07.4, OJ1548_F12.26, OSNPB_020805900
OJ1147_D11.1-1, OSNPB_060199200
Chromosome Location:
-
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
-
Assay ID Os03624200_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4331069
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015770166.1 XP_015625652.1 2 - 3 328 79
Gene Symbol: LOC4340405
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK065642.1 - 368 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Oryza sativa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0