Assay Details
Gene Symbol: LOC4331069 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770166.1 | XP_015625652.1 | 1 - 2 | 173 | 56 | |
GenBank mRNA | AK073740.1 | - | 1 - 2 | 179 | 56 |
Gene Symbol: LOC4340405 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK065642.1 | - | 523 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
