Assay Details
Gene Symbol: LOC4330921 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015768091.2 | XP_015623577.1 | 3 - 4 | 558 | 69 | |
GenBank mRNA | AK067608.1 | - | 3 - 4 | 516 | 69 | |
AK121526.1 | - | 3 - 4 | 559 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
