Assay Details
Gene Symbol: LOC4330749 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772108.2 | XP_015627594.1 | 5 - 6 | 798 | 152 | |
XM_015772109.2 | XP_015627595.1 | 5 - 6 | 800 | 152 | ||
GenBank mRNA | AK062267.1 | - | 5 - 6 | 731 | 152 | |
AK066184.1 | - | 5 - 6 | 735 | 152 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
