Assay Details
Gene Symbol: LOC9271572 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777004.2 | XP_015632490.1 | 2 - 3 | 526 | 67 | |
XM_015777012.2 | XP_015632498.1 | 2 - 3 | 526 | 67 | ||
GenBank mRNA | AK108568.1 | - | 2 - 3 | 532 | 67 | |
AY553724.1 | - | 1 - 2 | 305 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
