This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03620587_m1
See other LOC9271572 GE Assays ›
Gene Symbol:
LOC9271572
Gene Name:
MADS-box transcription factor 3-like
Gene Aliases:
MADS3, OSNPB_010201700, OsMADS3, RAG, RMADS222
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
Assay ID Os03620587_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC9271572
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015777004.2 XP_015632490.1 1 - 2 231 154
XM_015777012.2 XP_015632498.1 1 - 2 231 154
GenBank mRNA AK108568.1 - 1 - 2 237 154

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_015777004.2, XM_015777012.2 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Oryza sativa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0