Assay Details
Gene Symbol: LOC4348824 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015759148.1 | XP_015614634.1 | 7 - 8 | 918 | 149 | |
GenBank mRNA | AK069617.1 | - | 7 - 8 | 915 | 149 | |
AK119623.1 | - | 7 - 8 | 843 | 149 | ||
GU120332.1 | - | 7 - 8 | 794 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
