Assay Details
Gene Symbol: LOC4346999 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015757133.2 | XP_015612619.1 | 4 - 5 | 451 | 68 | |
XM_015757134.2 | XP_015612620.1 | 4 - 5 | 448 | 68 | ||
GenBank mRNA | AK063980.1 | - | 4 - 5 | 442 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
