This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03615786_g1
See other LOC4344973 GE Assays ›
Gene Symbol:
LOC4344973 LOC4345935
Gene Name:
3-hydroxyacyl-[acyl-carrier-protein] dehydratase FabZ
tricin synthase 2-like
Gene Aliases:
OSNPB_080225000, P0494D11.8
OSNPB_080498400, OsJ_27815, ROMT-17
Chromosome Location:
-
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
-
Assay ID Os03615786_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4344973
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK287940.1 - 1654 63
Gene Symbol: LOC4345935
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015794277.2 XP_015649763.1 2 - 3 324 63
GenBank mRNA AK061757.1 - 2 - 3 296 63
AK065515.1 - 1 - 2 217 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Oryza sativa Probe spans exons

Panther Classification:

Molecular Function -

methyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0