Assay Details
Gene Symbol: LOC4345611 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015793283.2 | XP_015648769.1 | 3 - 4 | 505 | 64 | |
GenBank mRNA | AK099569.1 | - | 3 - 4 | 483 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
