Assay Details
Gene Symbol: LOC4333983 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777179.2 | XP_015632665.1 | 11 - 12 | 1637 | 79 | |
GenBank mRNA | AK068068.1 | - | 11 - 12 | 1635 | 79 | |
AK121524.1 | - | 2 - 3 | 200 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
