Assay Details
Gene Symbol: LOC4330985 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770017.2 | XP_015625503.1 | 1 - 2 | 1215 | 74 | |
GenBank mRNA | AK121357.1 | - | 1 - 2 | 1204 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
