Assay Details
Gene Symbol: LOC4330930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771474.2 | XP_015626960.1 | 3 - 4 | 1397 | 126 | |
GenBank mRNA | AK065421.1 | - | 3 - 4 | 930 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
