Assay Details
Gene Symbol: LOC4352932 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015764222.2 | XP_015619708.1 | 2 - 3 | 779 | 80 | |
GenBank mRNA | AK065733.1 | - | 2 - 3 | 776 | 80 | |
AK120333.1 | - | 2 - 3 | 776 | 80 | ||
AK242597.1 | - | 2 - 3 | 776 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
