This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03599861_g1
See other LOC4351753 GE Assays ›
Gene Symbol:
LOC4351753
Gene Name:
MADS-box transcription factor 13-like
Gene Aliases:
MADS13, MAF1, OSNPB_120207000, OsMADS13, RMADS206
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
Assay ID Os03599861_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4351753
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015762996.2 XP_015618482.1 4 - 5 510 73
XM_015762997.2 XP_015618483.1 4 - 5 553 73
XM_015762998.2 XP_015618484.1 4 - 5 421 73
XM_015763000.2 XP_015618486.1 4 - 5 508 73
XM_015763001.2 XP_015618487.1 4 - 5 553 73
GenBank mRNA AK070425.1 - 4 - 5 404 73
AY332475.1 - 3 - 4 377 73
AY551912.1 - 3 - 4 486 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Oryza sativa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0