Assay Details
Gene Symbol: LOC4326986 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772911.2 | XP_015628397.1 | 2 - 3 | 875 | 65 | |
GenBank mRNA | AK120056.1 | - | 2 - 3 | 876 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
