This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03588533_m1
See other LOC4332947 GE Assays ›
Gene Symbol:
LOC4332947
Gene Name:
protein COP1 SUPPRESSOR 2
Gene Aliases:
OSNPB_030374100
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
Assay ID Os03588533_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4332947
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015772316.2 XP_015627802.1 5 - 6 828 102
GenBank mRNA AK066002.1 - 5 - 6 809 102
AK099328.1 - 5 - 6 727 102
AK104845.1 - 5 - 6 773 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Oryza sativa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0