Assay Details
Gene Symbol: LOC4332947 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772316.2 | XP_015627802.1 | 2 - 3 | 487 | 70 | |
GenBank mRNA | AK066002.1 | - | 2 - 3 | 468 | 70 | |
AK099328.1 | - | 2 - 3 | 386 | 70 | ||
AK104845.1 | - | 2 - 3 | 432 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
