This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03584850_m1
See other LOC4326908 GE Assays ›
Gene Symbol:
LOC4326908
Gene Name:
3'-5' exoribonuclease 1
Gene Aliases:
B1085F09.4, OSNPB_010280000
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
Assay ID Os03584850_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4326908
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015766150.2 XP_015621636.1 2 - 3 741 92
GenBank mRNA AK062026.1 - 1 - 2 323 92
AK103348.1 - 2 - 3 739 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Oryza sativa Probe spans exons

Panther Classification:

Molecular Function -

exoribonuclease

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0