This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03584565_g1
See other LOC4326296 GE Assays ›
Gene Symbol:
LOC4326296
Gene Name:
light-mediated development protein DET1
Gene Aliases:
OSNPB_010104600
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
Assay ID Os03584565_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4326296
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015766621.2 XP_015622107.1 10 - 11 1836 108
XM_015766622.2 XP_015622108.1 10 - 11 1833 108
GenBank mRNA AK058683.1 - 10 - 11 1532 108
AK100613.1 - 10 - 11 1835 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Oryza sativa Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase protein modifying enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0